Assay Details
| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008957.4 | XP_004009006.2 | 3 - 4 | 980 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL17B| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008957.4 | XP_004009006.2 | 3 - 4 | 980 | 72 | |
Set Membership: |
|