Assay Details
Gene Symbol: RIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010893.4 | XP_004010942.2 | 7 - 8 | 1062 | 64 | |
XM_012180911.3 | XP_012036301.2 | 7 - 8 | 1059 | 64 | ||
XM_012180912.3 | XP_012036302.2 | 6 - 7 | 921 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
