Assay Details
Gene Symbol: LOC101121723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019389.4 | XP_004019438.2 | 15 - 16 | 1764 | 131 | |
XM_012101586.3 | XP_011956976.2 | 15 - 16 | 1755 | 131 |
Gene Symbol: LOC114110156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960063.1 | XP_027815864.1 | 2 - 3 | 206 | 131 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
