Assay Details
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012094732.3 | XP_011950122.2 | 6 - 7 | 898 | 79 | |
XM_012094753.2 | XP_011950143.2 | 4 - 5 | 542 | 79 | ||
XM_027980069.1 | XP_027835870.1 | 6 - 7 | 896 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
