Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096771.3 | XP_011952161.1 | 2 - 3 | 795 | 85 | |
XM_015101251.2 | XP_014956737.1 | 2 - 3 | 743 | 85 | ||
XM_027980012.1 | XP_027835813.1 | 1 - 2 | 2710 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
