Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 17 - 18 | 5073 | 79 | |
| XM_012098428.3 | XP_011953818.2 | 12 - 13 | 3057 | 79 | ||
| XM_027975368.1 | XP_027831169.1 | 21 - 22 | 5780 | 79 | ||
| XM_027975369.1 | XP_027831170.1 | 21 - 22 | 5780 | 79 | ||
| XM_027975370.1 | XP_027831171.1 | 21 - 22 | 5780 | 79 | ||
| XM_027975371.1 | XP_027831172.1 | 21 - 22 | 5780 | 79 | ||
| XM_027975372.1 | XP_027831173.1 | 20 - 21 | 5714 | 79 | ||
| XM_027975373.1 | XP_027831174.1 | 19 - 20 | 5660 | 79 | ||

LOC101115486
Genomic Map