Assay Details
| Gene Symbol: GPSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100905.3 | XP_011956295.1 | 3 - 4 | 707 | 61 | |
| XM_015102774.2 | XP_014958260.1 | 3 - 4 | 736 | 61 | ||
More Information
Set Membership: |
|

GPSM3
Genomic Map