Assay Details
Gene Symbol: LOC101116577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182163.3 | XP_012037553.1 | 3 - 4 | 1379 | 97 | |
XM_027972087.1 | XP_027827888.1 | 3 - 4 | 1379 | 97 | ||
XM_027972088.1 | XP_027827889.1 | 12 - 13 | 3750 | 97 | ||
XM_027972089.1 | XP_027827890.1 | 3 - 4 | 1379 | 97 |
