Assay Details
| Gene Symbol: ASIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098503.2 | XP_014953989.1 | 9 - 10 | 1839 | 71 | |
| XM_027975288.1 | XP_027831089.1 | 9 - 10 | 1597 | 71 | ||
More Information
Set Membership: |
|

ASIC2
Genomic Map