Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098735.2 | XP_014954221.2 | 5 - 6 | 1616 | 70 | |
XM_027974650.1 | XP_027830451.1 | 5 - 6 | 1600 | 70 | ||
XM_027974651.1 | XP_027830452.1 | 5 - 6 | 1622 | 70 | ||
XM_027974652.1 | XP_027830453.1 | 5 - 6 | 1568 | 70 | ||
XM_027974653.1 | XP_027830454.1 | 4 - 5 | 1510 | 70 | ||
XM_027974654.1 | XP_027830455.1 | 4 - 5 | 1482 | 70 | ||
XM_027974655.1 | XP_027830456.1 | 4 - 5 | 1478 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
