Assay Details
Gene Symbol: LOC101102057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008561.4 | XP_004008610.1 | 1 - 2 | 107 | 79 | |
XM_027969726.1 | XP_027825527.1 | 1 - 2 | 107 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
