Assay Details
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958650.1 | XP_027814451.1 | 4 - 5 | 1406 | 78 | |
XM_027958651.1 | XP_027814452.1 | 4 - 5 | 1403 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
