Assay Details
| Gene Symbol: HPRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015105023.2 | XP_014960509.1 | 7 - 8 | 686 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPRT1| Gene Symbol: HPRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015105023.2 | XP_014960509.1 | 7 - 8 | 686 | 89 | |
Set Membership: |
|