Assay Details
| Gene Symbol: REM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181006.3 | XP_012036396.3 | 3 - 4 | 3376 | 79 | |
| XM_027971647.1 | XP_027827448.1 | 3 - 4 | 3379 | 79 | ||
More Information
Set Membership: |
|

REM2
Genomic Map