Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093486.2 | XP_014948972.1 | 1 - 2 | 338 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASP10| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093486.2 | XP_014948972.1 | 1 - 2 | 338 | 52 | |
Set Membership: |
|