Assay Details
| Gene Symbol: LOC101103275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958587.1 | XP_027814388.1 | 5 - 6 | 598 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103275| Gene Symbol: LOC101103275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958587.1 | XP_027814388.1 | 5 - 6 | 598 | 81 | |
Set Membership: |
|