Assay Details
| Gene Symbol: GTF2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980298.1 | XP_027836099.1 | 4 - 5 | 1108 | 90 | |
| XM_027980303.1 | XP_027836104.1 | 4 - 5 | 1078 | 90 | ||
| XM_027980311.1 | XP_027836112.1 | 4 - 5 | 1069 | 90 | ||
| XM_027980320.1 | XP_027836121.1 | 4 - 5 | 1283 | 90 | ||
More Information
Set Membership: |
|

GTF2E1
Genomic Map