Assay Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012142189.2 | XP_011997579.1 | 6 - 7 | 1027 | 63 | |
XM_015093483.2 | XP_014948969.1 | 4 - 5 | 966 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
