Assay Details
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103598.2 | XP_014959084.1 | 2 - 3 | 218 | 76 | |
XM_027960718.1 | XP_027816519.1 | 2 - 3 | 441 | 76 | ||
XM_027960719.1 | XP_027816520.1 | 3 - 4 | 564 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
