Assay Details
Gene Symbol: LOC101113488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094104.2 | XP_014949590.2 | 5 - 6 | 535 | 80 | |
XM_015094105.2 | XP_014949591.1 | 2 - 3 | 258 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
