Assay Details
| Gene Symbol: DUSP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962180.1 | XP_027817981.1 | 2 - 3 | 319 | 66 | |
| XM_027962181.1 | XP_027817982.1 | 2 - 3 | 326 | 66 | ||
More Information
Set Membership: |
|

DUSP29
Genomic Map