Assay Details
Gene Symbol: SPG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977759.1 | XP_027833560.1 | 4 - 5 | 643 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
