Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004265.4 | XP_004004314.1 | 3 - 4 | 397 | 78 | |
XM_004004266.4 | XP_004004315.1 | 3 - 4 | 634 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
