Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014924.3 | XP_004014973.1 | 3 - 4 | 349 | 94 | |
| XM_027977651.1 | XP_027833452.1 | 3 - 4 | 263 | 94 | ||
More Information
Set Membership: |
|

DYNLRB2
Genomic Map