Assay Details
| Gene Symbol: APCDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103585.3 | XP_011958975.2 | 4 - 5 | 1254 | 65 | |
| XM_012103586.3 | XP_011958976.2 | 4 - 5 | 1586 | 65 | ||
More Information
Set Membership: |
|

APCDD1
Genomic Map