Assay Details
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974210.1 | XP_027830011.1 | 4 - 5 | 544 | 63 | |
XM_027974211.1 | XP_027830012.1 | 4 - 5 | 535 | 63 | ||
XM_027974212.1 | XP_027830013.1 | 3 - 4 | 342 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
