Assay Details
Gene Symbol: LOC101110111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967129.1 | XP_027822930.1 | 3 - 4 | 384 | 73 | |
XM_027967134.1 | XP_027822935.1 | 3 - 4 | 344 | 73 | ||
XM_027967140.1 | XP_027822941.1 | 3 - 4 | 287 | 73 | ||
XM_027967147.1 | XP_027822948.1 | 3 - 4 | 329 | 73 | ||
XM_027967151.1 | XP_027822952.1 | 3 - 4 | 318 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
