Assay Details
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002907.4 | XP_004002956.1 | 2 - 3 | 1174 | 64 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002907.4 | XP_004002956.1 | 2 - 3 | 1174 | 64 |