Assay Details
| Gene Symbol: LOC101118051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016905.4 | XP_004016954.1 | 13 - 14 | 1494 | 117 | |
| XM_012096792.3 | XP_011952182.1 | 14 - 15 | 1587 | 117 | ||
| XM_027980025.1 | XP_027835826.1 | 12 - 13 | 1916 | 117 | ||
| XM_027980026.1 | XP_027835827.1 | 13 - 14 | 1952 | 117 | ||
More Information
Set Membership: |
|

LOC101118051
Genomic Map