Assay Details
| Gene Symbol: LOC101117706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018513.3 | XP_004018562.2 | 2 - 3 | 1240 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117706| Gene Symbol: LOC101117706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018513.3 | XP_004018562.2 | 2 - 3 | 1240 | 68 | |
Set Membership: |
|