Assay Details
| Gene Symbol: TREX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023398.4 | XP_004023447.4 | 1 - 2 | 585 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004023398.4 transcripts.
Set Membership: |
|

TREX1
Genomic Map