Assay Details
| Gene Symbol: C5H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177809.3 | XP_012033199.2 | 2 - 3 | 133 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012177809.3 transcripts.
Set Membership: |
|

C5H19orf57
Genomic Map