Assay Details
Gene Symbol: C5H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177809.3 | XP_012033199.2 | 3 - 4 | 315 | 89 | |
XM_027969515.1 | XP_027825316.1 | 3 - 4 | 568 | 89 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177809.3 | XP_012033199.2 | 3 - 4 | 315 | 89 | |
XM_027969515.1 | XP_027825316.1 | 3 - 4 | 568 | 89 |