Assay Details
| Gene Symbol: LOC101116193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003245.3 | XP_004003294.1 | 1 - 2 | 136 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101116193| Gene Symbol: LOC101116193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003245.3 | XP_004003294.1 | 1 - 2 | 136 | 92 | |
Set Membership: |
|