Assay Details
Gene Symbol: LOC101116193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003245.3 | XP_004003294.1 | 2 - 3 | 364 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
