Assay Details
Gene Symbol: RBM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017180.4 | XP_004017229.1 | 3 - 4 | 694 | 72 | |
XM_027980317.1 | XP_027836118.1 | 2 - 3 | 623 | 72 | ||
XM_027980318.1 | XP_027836119.1 | 2 - 3 | 623 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
