Assay Details
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021000.3 | XP_004021049.3 | 3 - 4 | 2013 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
