Assay Details
Gene Symbol: SAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104542.3 | XP_011959932.1 | 2 - 3 | 579 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
