Assay Details
Gene Symbol: LOC114112283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963774.1 | XP_027819575.1 | 3 - 4 | 375 | 93 |
Gene Symbol: LOC101121916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963210.1 | XP_027819011.1 | 7 - 8 | 974 | 93 |
