Assay Details
Gene Symbol: LOC101121661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012129529.3 | XP_011984919.2 | 8 - 9 | 1069 | 104 | |
XM_012129535.2 | XP_011984925.2 | 8 - 9 | 864 | 104 | ||
XM_027964634.1 | XP_027820435.1 | 8 - 9 | 1073 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
