Assay Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094908.2 | XP_014950394.2 | 3 - 4 | 1002 | 88 | |
| XM_027967831.1 | XP_027823632.1 | 3 - 4 | 459 | 88 | ||
More Information
Set Membership: |
|

AEBP2
Genomic Map