Assay Details
| Gene Symbol: LOC101113122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976731.1 | XP_027832532.1 | 7 - 8 | 1852 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113122| Gene Symbol: LOC101113122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976731.1 | XP_027832532.1 | 7 - 8 | 1852 | 119 | |
Set Membership: |
|