Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011435.4 | XP_004011484.3 | 6 - 7 | 616 | 67 | |
XM_027972629.1 | XP_027828430.1 | 6 - 7 | 514 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
