Assay Details
| Gene Symbol: KIAA0513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014947.3 | XP_004014996.1 | 1 - 2 | 604 | 58 | |
| XM_012189643.2 | XP_012045033.1 | 2 - 3 | 553 | 58 | ||
| XM_027977693.1 | XP_027833494.1 | 2 - 3 | 615 | 58 | ||
More Information
Set Membership: |
|

KIAA0513
Genomic Map