Assay Details
Gene Symbol: CLDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020967.4 | XP_004021016.1 | 1 | 772 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
