Assay Details
| Gene Symbol: SLC39A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973448.1 | XP_027829249.1 | 5 - 6 | 1098 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A4| Gene Symbol: SLC39A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973448.1 | XP_027829249.1 | 5 - 6 | 1098 | 57 | |
Set Membership: |
|