Assay Details
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965801.1 | XP_027821602.1 | 1 - 2 | 224 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
