Assay Details
Gene Symbol: LOC101104231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008016.4 | XP_004008065.1 | 11 - 12 | 2127 | 109 | |
XM_004008018.4 | XP_004008067.1 | 11 - 12 | 2127 | 109 | ||
XM_012177044.3 | XP_012032434.1 | 11 - 12 | 2128 | 109 | ||
XM_012177047.3 | XP_012032437.1 | 11 - 12 | 2128 | 109 | ||
XM_012177052.3 | XP_012032442.1 | 11 - 12 | 2127 | 109 | ||
XM_012177054.3 | XP_012032444.1 | 11 - 12 | 2127 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
