Assay Details
Gene Symbol: LOC101104231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008016.4 | XP_004008065.1 | 16 - 17 | 2745 | 93 | |
XM_004008017.4 | XP_004008066.1 | 17 - 18 | 2755 | 93 | ||
XM_012177043.3 | XP_012032433.1 | 17 - 18 | 2755 | 93 | ||
XM_012177047.3 | XP_012032437.1 | 16 - 17 | 2746 | 93 | ||
XM_012177054.3 | XP_012032444.1 | 16 - 17 | 2745 | 93 | ||
XM_027968767.1 | XP_027824568.1 | 17 - 18 | 2754 | 93 | ||
XM_027968768.1 | XP_027824569.1 | 17 - 18 | 2754 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
