Assay Details
Gene Symbol: LOC101104231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008016.4 | XP_004008065.1 | 1 - 2 | 601 | 79 | |
XM_004008017.4 | XP_004008066.1 | 1 - 2 | 602 | 79 | ||
XM_004008018.4 | XP_004008067.1 | 1 - 2 | 601 | 79 | ||
XM_004008019.4 | XP_004008068.1 | 1 - 2 | 602 | 79 | ||
XM_012177037.3 | XP_012032427.1 | 1 - 2 | 602 | 79 | ||
XM_012177040.3 | XP_012032430.1 | 1 - 2 | 602 | 79 | ||
XM_012177041.3 | XP_012032431.1 | 1 - 2 | 602 | 79 | ||
XM_012177042.3 | XP_012032432.1 | 1 - 2 | 603 | 79 | ||
XM_012177043.3 | XP_012032433.1 | 1 - 2 | 602 | 79 | ||
XM_012177044.3 | XP_012032434.1 | 1 - 2 | 602 | 79 | ||
XM_012177046.3 | XP_012032436.1 | 1 - 2 | 604 | 79 | ||
XM_012177047.3 | XP_012032437.1 | 1 - 2 | 602 | 79 | ||
XM_012177049.3 | XP_012032439.1 | 1 - 2 | 601 | 79 | ||
XM_012177051.3 | XP_012032441.1 | 1 - 2 | 606 | 79 | ||
XM_012177052.3 | XP_012032442.1 | 1 - 2 | 601 | 79 | ||
XM_012177053.3 | XP_012032443.1 | 1 - 2 | 601 | 79 | ||
XM_012177054.3 | XP_012032444.1 | 1 - 2 | 601 | 79 | ||
XM_027968766.1 | XP_027824567.1 | 1 - 2 | 601 | 79 | ||
XM_027968767.1 | XP_027824568.1 | 1 - 2 | 601 | 79 | ||
XM_027968768.1 | XP_027824569.1 | 1 - 2 | 601 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
